missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride, Certified, 0.1000N ±0.0005N (0.1M), LabChem™

Supplier:  LabChem LC235702

 View more versions of this product

Catalog No. LC235702


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium Chloride
Colorless
Liquid
ClNa
MFCD00003477
Soluble in water
[Na+].[Cl-]
58.44
CHEBI:26710
Certified
Passes Test
Hydrogen chloride
7647-14-5,7732-18-5
99.42,0.58
1 L
NaCl
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
FAPWRFPIFSIZLT-UHFFFAOYSA-M
sodium chloride
5234
58.44
0.1000N ±0.0005N (0.1M)
Poly Bottle

Chemical Identifiers

7647-14-5
58.44
FAPWRFPIFSIZLT-UHFFFAOYSA-M
5234
sodium chloride
ClNa
MFCD00003477
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
CHEBI:26710
[Na+].[Cl-]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions