missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium chloride, ACS Reagent, Honeywell Fluka™
ACS Reagent,≥99.0%
$133.15 - $1767.90
Chemical Identifiers
| CAS | 7647-14-5 |
|---|---|
| Molecular Formula | ClNa |
| Molecular Weight (g/mol) | 58.44 |
| MDL Number | MFCD00003477 |
| InChI Key | FAPWRFPIFSIZLT-UHFFFAOYSA-M |
| Synonym | sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex |
| PubChem CID | 5234 |
| ChEBI | CHEBI:26710 |
| SMILES | [Na+].[Cl-] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
6003219
|
Honeywell Chemicals
S9888500G |
500 g |
Each for $133.15
|
|
|||||
|
6003213
|
Honeywell Chemicals
S98881KG |
1 kg |
Each for $211.38
|
|
|||||
|
6003214
|
Honeywell Chemicals
S988825KGDUP |
2.5 kg |
Each for $651.20
|
|
|||||
|
6003222
|
Honeywell Chemicals
S98886X500G |
6 x 500 g |
Case of 6 Each for $719.83
|
|
|||||
|
6003221
|
Honeywell Chemicals
S98885KG |
5 kg |
Each for $581.75
|
|
|||||
|
6003218
|
Honeywell Chemicals
S98884X25KG |
4 x 2.5 kg |
Case of 4 Each for $983.55
|
|
|||||
|
6003212
|
Honeywell Chemicals
S988810KG |
10 kg |
Each for $630.65
|
|
|||||
|
6003215
|
Honeywell Chemicals
S988820KG |
20 kg |
Each for $1,277.53
|
|
|||||
|
6003220
|
Honeywell Chemicals
S988850KG |
50 kg |
Each for $1,767.90
|
|
|||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 7647-14-5 | |
| 58.44 | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| 5234 | |
| [Na+].[Cl-] |
| ClNa | |
| MFCD00003477 | |
| sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
| CHEBI:26710 |
Specifications
| 7647-14-5 | |
| ClNa | |
| MFCD00003477 | |
| 3534976 | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| sodium chloride | |
| 5234 | |
| 58.44g/mol | |
| ACS Reagent |
| 500 g | |
| NaCl | |
| NONH for all modes of transport | |
| sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
| [Na+].[Cl-] | |
| 58.44 | |
| CHEBI:26710 | |
| ≥99% | |
| Sodium chloride |
Safety and Handling
RTECSNumber : VZ4725000