missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride, ACS Reagent Grade, Ricca Chemical

Catalog No. RDCS0430500
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDCS0430500 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RDCS0430500 Supplier Ricca Chemical Company Supplier No. RDCS0430500B1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
ChEBI CHEBI:26710
IUPAC Name sodium chloride

Specifications

Color White
CAS Min % 98.0
CAS Max % 100.0
Physical Form Powder
Quantity 500 g
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
SMILES [Na+].[Cl-]
PubChem CID 5234
Percent Purity ≥ 99%
Grade ACS
Packaging White Poly Wide Mouth Bottle
Chemical Name or Material Sodium Chloride
Show More Show Less