Learn More
Sodium Chloride, ACS, ≥99%, Ultrapure
Used for precipitation of DNA | CAS: 7647-14-5 | ClNa | 58.44 g/mol
Supplier: Thermo Fisher Scientific J21618A7
Description
Used for the precipitation of DNA.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 1413°C | |
| 801°C | |
| Crystalline Powder | |
| ClNa | |
| sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
| [Na+].[Cl-] | |
| 58.44 | |
| CHEBI:26710 | |
| ≥99% | |
| Plastic bottle | |
| 2.165 |
| Sodium Chloride | |
| 7647-14-5 | |
| 5 kg | |
| MFCD00003477 | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| sodium chloride | |
| 5234 | |
| 58.44 | |
| ACS | |
| 2.165 |
Chemical Identifiers
| 7647-14-5 | |
| 58.44 | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| 5234 | |
| [Na+].[Cl-] |
| ClNa | |
| MFCD00003477 | |
| sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
| CHEBI:26710 |
Safety and Handling
EINECSNumber : 231-598-3
RTECSNumber : VZ4725000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only