missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride, ACS, ≥99%, Ultrapure

Catalog No. AAJ21618A9
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
10 kg
Packaging:
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ21618A9 10 kg Plastic bottle
AAJ2161836 500 g Plastic bottle
AAJ21618A1 1 kg Plastic bottle
AAJ21618A7 5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAJ21618A9 Supplier Thermo Fisher Scientific Supplier No. J21618A9

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used for precipitation of DNA | CAS: 7647-14-5 | ClNa | 58.44 g/mol

Used for the precipitation of DNA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
SMILES [Na+].[Cl-]

Specifications

Boiling Point 1413°C
Melting Point 801°C
Physical Form Crystalline Powder
Quantity 10 kg
IUPAC Name sodium;chloride
Formula Weight 58.44
Percent Purity ≥99%
Grade ACS
Packaging Plastic bottle
Specific Gravity 2.165
Density 2.165
Chemical Name or Material Sodium Chloride
Show More Show Less
EINECSNumber 231-598-3
RTECSNumber VZ4725000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only