missing translation for 'onlineSavingsMsg'
Learn More

Sodium chloride, 99+%, ACS reagent

CAS: 7647-14-5 | ClNa | 58.44 g/mol

Supplier:  Thermo Scientific Chemicals 424290050

 View more versions of this product

Catalog No. AC424290050


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1461.0°C
801.0°C
Brown-Yellow to Orange
Adhering Powder and/or Lumps
99+%
NaCl
13,280
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
FAPWRFPIFSIZLT-UHFFFAOYSA-M
sodium chloride
5234
58.44
ACS Reagent
≤0.5%
1mmHg at 440°C
Sodium chloride
7647-14-5
5.0 to 9.0 (5% soln. at 25°C)
5 kg
ClNa
MFCD00003477
15, 8734
Solubility in water: 360g/L (20°C). Other solubilities: soluble in glycerol (1g/10mL), ammonia, slightly soluble in alcohol, nearly insoluble in hydrochloric acid
[Na+].[Cl-]
58.44
CHEBI:26710
≥99%
Pass Test
Amber Glass Bottle

Chemical Identifiers

7647-14-5
58.44
FAPWRFPIFSIZLT-UHFFFAOYSA-M
5234
[Na+].[Cl-]
ClNa
MFCD00003477
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
CHEBI:26710
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only