missing translation for 'onlineSavingsMsg'
Learn More

Sodium chloride, 99+%, ACS reagent

Catalog No. AC424290250
Change view
Click to view available options
Quantity:
25 g
500 g
1 kg
5 kg
25 kg
Packaging:
Amber Glass Bottle
Fibre drum
Plastic Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC424290250 25 g Plastic Bottle
AC424295000 500 g Amber Glass Bottle
AC424290010 1 kg Amber Glass Bottle
AC424290050 5 kg Amber Glass Bottle
AC424290251 25 kg Fibre drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AC424290250 Supplier Thermo Scientific Chemicals Supplier No. 424290250

CAS: 7647-14-5 | ClNa | 58.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
SMILES [Na+].[Cl-]

Specifications

Boiling Point 1461.0°C
Melting Point 801.0°C
Color Brown-Yellow to Orange
pH 5.0 to 9.0 (5% soln. at 25°C)
Physical Form Adhering Powder and/or Lumps
Quantity 25 g
Assay Percent Range 99% min. (Argentometry)
Linear Formula NaCl
Fieser 13,280
Merck Index 15, 8734
Solubility Information Solubility in water: 360g/L (20°C). Other solubilities: soluble in glycerol (1g/10mL), ammonia, slightly soluble in alcohol, nearly insoluble in hydrochloric acid
IUPAC Name sodium chloride
Formula Weight 58.44
Percent Purity ≥99%
Grade ACS Reagent
Identification Pass Test
Loss on Drying ≤0.5%
Packaging Plastic Bottle
Vapor Pressure 1mmHg at 440°C
Chemical Name or Material Sodium chloride
Show More Show Less
EINECSNumber 231-598-3

RUO – Research Use Only