missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium chloride, 99.99%, Suprapur™, MilliporeSigma™
$1301.69 - $1301.69
Chemical Identifiers
| CAS | 7647-14-5 |
|---|---|
| Molecular Formula | ClNa |
| Molecular Weight (g/mol) | 58.44 |
| MDL Number | MFCD00003477 |
| InChI Key | FAPWRFPIFSIZLT-UHFFFAOYSA-M |
| Synonym | sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex |
| PubChem CID | 5234 |
| ChEBI | CHEBI:26710 |
| IUPAC Name | sodium chloride |
| SMILES | [Na+].[Cl-] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
M1064060500
|
Technidata
1064060500 |
500 g | Plastic Bottle |
Each for $1,301.69
|
|
||||
Chemical Identifiers
| 7647-14-5 | |
| 58.44 | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| 5234 | |
| sodium chloride |
| ClNa | |
| MFCD00003477 | |
| sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
| CHEBI:26710 | |
| [Na+].[Cl-] |
Specifications
| Colorless | |
| Solid | |
| 99.99% | |
| NaCl | |
| sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| sodium chloride | |
| 5234 | |
| 58.44 | |
| Plastic Bottle | |
| 2.17g/cm3 (20°C) |
| 7647-14-5 | |
| 500 g | |
| ClNa | |
| MFCD00003477 | |
| 358g/L (20°C) | |
| [Na+].[Cl-] | |
| 58.44 | |
| CHEBI:26710 | |
| Suprapur | |
| 1.3 hPa (865°C) | |
| Sodium chloride |