missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride, 99.5% min., (USP), MP Biomedicals

Catalog No. p-3547309
Change view
Click to view available options
Quantity:
500 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN1028925 500 g
ICN10289201 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN1028925 Supplier MP Biomedicals Inc Supplier No. 02102892.5
Only null left

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Quantity 500 g
Assay Percent Range 99.5% min.
Formula Weight 58.44
Grade USP
Chemical Name or Material Sodium Chloride
Recommended Storage Room Temperature