missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Chloride, 99.5% min., (USP), MP Biomedicals
Specifications
Specifications
| Quantity | 500 g |
| Assay Percent Range | 99.5% min. |
| Formula Weight | 58.44 |
| Grade | USP |
| Chemical Name or Material | Sodium Chloride |
Safety and Handling
| Recommended Storage | Room Temperature |