missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride 5M, Corning™

Catalog No. MT46032CV
Change view
Click to view available options
Quantity:
3 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MT46032CV 3 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MT46032CV Supplier Corning Incorporated Supplier No. 46032CV

Specifications

Form Liquid
Product Type Sodium Chloride
Molecular Formula ClNa
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
SMILES [Na+].[Cl-]
IUPAC Name sodium chloride
Molecular Weight (g/mol) 58.44
Quantity 3 L