missing translation for 'onlineSavingsMsg'
Learn More

Sodium carbonate/Sodium hydroxide, IC eluent, Concentrate (20x) for Metrosep A Supp 16, MilliporeSigma™ Supelco™

This eluent concentrate is application tested. It is certified in accordance with ISO Guide 31. All details about exact content, uncertainty, traceability and expiry date are described in the certificate.

$347.78 - $844.92

Specifications

CAS 497-19-8
Concentration or Composition (by Analyte or Components) 150 mM Na2 CO3 aq. soln.
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111002616
View Documents
Merck Emd Millipore
383021L
1 L N/A
Only null left
 
111002614
View Documents
Merck Emd Millipore
3830225L
2.5 L N/A
Only null left
 
Description

Description

Specifications

Specifications

497-19-8
150 mM Na2 CO3 aq. soln.
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'