missing translation for 'onlineSavingsMsg'
Learn More

Sodium carbonate/Sodium hydroxide, IC eluent, Concentrate (20x) for Metrosep A Supp 16, MilliporeSigma™ Supelco™

Catalog No. 111002616
Change view
Click to view available options
Quantity:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002616 1 L
111002614 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111002616 Supplier Merck Emd Millipore Supplier No. 383021L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This eluent concentrate is application tested. It is certified in accordance with ISO Guide 31. All details about exact content, uncertainty, traceability and expiry date are described in the certificate.

Specifications

CAS 497-19-8
Physical Form Liquid
Quantity 1 L
Concentration or Composition (by Analyte or Components) 150 mM Na2 CO3 aq. soln.