missing translation for 'onlineSavingsMsg'
Learn More

Sodium carbonate, Certified by BAM, ≥99.5%, MilliporeSigma™ Supelco™

Catalog No. 111014323
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014323 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014323 Supplier Merck Emd Millipore Supplier No. 7136350G

Specifications

CAS 497-19-8
Physical Form Neat
Linear Formula Na2 CO3
MDL Number MFCD00003494
Melting Point 851°C
Synonym Calcined soda; Carbonic acid disodium salt; Soda ash
Type Certified Reference Material
Quantity 50 g
Formula Weight 105.99
Percent Purity ≥99.5%
Grade Certified Reference Material
Show More Show Less