missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Carbonate, Certified, 5.0% (w/v) ±0.5%, LabChem™
Specifications
Specifications
| Color | Colorless |
| CAS Max % | 95, 5 |
| Physical Form | Liquid |
| Quantity | 1 L |
| Linear Formula | Na2CO3 |
| Solubility Information | Soluble in water |
| Formula Weight | 105.99 |
| Percent Purity | 5.0% ±0.5% (w/v) |
| Grade | Certified |
| Identification | Passes Test |
| Show More |
Safety and Handling
| Hazard Statement | GHS H Statement Solution is not hazardous. |
| Precautionary Statement | GHS P Statement If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes. If swallowed, get medical attention. |
| Recommended Storage | Room Temperature |