missing translation for 'onlineSavingsMsg'
Learn More

Sodium Carbonate, Certified, 10.0% (w/v) ±0.5%, LabChem™

Catalog No. LC229801
Change view
Click to view available options
Quantity:
500 mL
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC229801 500 mL
LC229804 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC229801 Supplier LabChem Supplier No. LC229801
Only null left

Chemical Identifiers

CAS 497-19-8 , 7732-18-5
Molecular Formula CNa2O3
Molecular Weight (g/mol) 105.99
MDL Number MFCD00003494
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
Synonym sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
PubChem CID 10340
ChEBI CHEBI:29377
IUPAC Name disodium carbonate
SMILES [Na+].[Na+].[O-]C([O-])=O

Specifications

Color Colorless
CAS Max % 90, 10
Physical Form Liquid
Quantity 500 mL
Linear Formula Na2CO3
Solubility Information Soluble in water
Formula Weight 105.99
Percent Purity 10.0% ±0.5% (w/v)
Grade Certified
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1g/mL
Decomposition Information Fume; Carbon monoxide; Carbon dioxide
Density 1g/mL
Chemical Name or Material Sodium Carbonate
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Recommended Storage Room Temperature