missing translation for 'onlineSavingsMsg'
Learn More

Sodium Carbonate, Certified, 1.00N ±0.02N (0.5M), LabChem™

Catalog No. p-5388232
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Catalog No. Quantity
LC233801 500 mL
LC233802 1 L
LC233804 4 L
3 options

Catalog No. LC233801

Supplier: LabChem LC233801

Only null left

Chemical Identifiers

CAS 497-19-8 , 7732-18-5
Molecular Formula CNa2O3
Molecular Weight (g/mol) 105.99
MDL Number MFCD00003494
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
Synonym sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
PubChem CID 10340
ChEBI CHEBI:29377
IUPAC Name disodium carbonate
SMILES [Na+].[Na+].[O-]C([O-])=O

Specifications

Color Colorless
CAS Max % 94.7, 5.3
Physical Form Liquid
Quantity 500 mL
Linear Formula Na2CO3
Solubility Information Soluble in water
Formula Weight 105.99
Grade Certified
Concentration or Composition (by Analyte or Components) 1.00N ±0.02N (0.5M)
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1g/mL
Decomposition Information Fume; Carbon monoxide; Carbon dioxide
Density 1g/mL
Chemical Name or Material Sodium Carbonate
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature