missing translation for 'onlineSavingsMsg'
Learn More

Sodium Carbonate, Certified, 1.00N ±0.02N (0.5M), LabChem™

Supplier:  LabChem LC233802

 View more versions of this product

Catalog No. LC233802

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium Carbonate
Colorless
Liquid
CNa2O3
MFCD00003494
Soluble in water
[Na+].[Na+].[O-]C([O-])=O
105.99
CHEBI:29377
Certified
Passes Test
1g/mL
1g/mL
497-19-8,7732-18-5
94.7,5.3
1 L
Na2CO3
sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
CDBYLPFSWZWCQE-UHFFFAOYSA-L
disodium carbonate
10340
105.99
1.00N ±0.02N (0.5M)
Poly Bottle
Fume; Carbon monoxide; Carbon dioxide

Chemical Identifiers

497-19-8
105.99
CDBYLPFSWZWCQE-UHFFFAOYSA-L
10340
disodium carbonate
CNa2O3
MFCD00003494
sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
CHEBI:29377
[Na+].[Na+].[O-]C([O-])=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions