missing translation for 'onlineSavingsMsg'
Learn More

Sodium Carbonate, Certified, 0.530% (w/v) ±0.005%, LabChem™

Catalog No. lc229752
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 L
Packaging:
Poly Bottle
Catalog No. Quantity Packaging
LC229752 1 L Poly Bottle
1 options

Catalog No. LC229752

Supplier: LabChem LC229752

Only null left

Chemical Identifiers

CAS 497-19-8 , 7732-18-5
Molecular Formula CNa2O3
Molecular Weight (g/mol) 105.99
MDL Number MFCD00003494
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
Synonym sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
PubChem CID 10340
ChEBI CHEBI:29377
IUPAC Name disodium carbonate
SMILES [Na+].[Na+].[O-]C([O-])=O

Specifications

Color Colorless
CAS Max % 99.47, 0.53
Physical Form Liquid
Quantity 1 L
Linear Formula Na2CO3
Solubility Information Soluble in water
Formula Weight 105.99
Percent Purity 0.530% ± 0.005% (w/v)
Grade Certified
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1g/mL
Decomposition Information Fume; Carbon monoxide; Carbon dioxide
Density 1g/mL
Chemical Name or Material Sodium Carbonate
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature