missing translation for 'onlineSavingsMsg'
Learn More

Sodium Carbonate, Certified, 0.1000N ±0.0005N (0.05M), LabChem™

Catalog No. p-5388229
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC233501 500 mL
LC233502 1 L
LC233504 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC233501 Supplier LabChem Supplier No. LC233501
Only null left

Chemical Identifiers

CAS 497-19-8 , 7732-18-5
Molecular Formula CNa2O3
Molecular Weight (g/mol) 105.99
MDL Number MFCD00003494
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
Synonym sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
PubChem CID 10340
ChEBI CHEBI:29377
IUPAC Name disodium carbonate
SMILES [Na+].[Na+].[O-]C([O-])=O

Specifications

Color Colorless
CAS Max % 99.47, 0.53
Physical Form Liquid
Quantity 500 mL
Linear Formula Na2CO3
Solubility Information Soluble in water
Formula Weight 105.99
Grade Certified
Concentration or Composition (by Analyte or Components) 0.1000N ±0.0005N (0.05M)
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1g/mL
Decomposition Information Fume; Carbon monoxide; Carbon dioxide
Density 1g/mL
Chemical Name or Material Sodium Carbonate
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature