missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Carbonate, Certified, 0.1000N ±0.0005N (0.05M), LabChem™
Supplier: LabChem LC233504
Specifications
| Sodium Carbonate | |
| Colorless | |
| Liquid | |
| CNa2O3 | |
| MFCD00003494 | |
| Soluble in water | |
| [Na+].[Na+].[O-]C([O-])=O | |
| 105.99 | |
| CHEBI:29377 | |
| Certified | |
| Passes Test | |
| 1g/mL | |
| 1g/mL |
| 497-19-8,7732-18-5 | |
| 99.47,0.53 | |
| 4 L | |
| Na2CO3 | |
| sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda | |
| CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
| disodium carbonate | |
| 10340 | |
| 105.99 | |
| 0.1000N ±0.0005N (0.05M) | |
| Poly Bottle | |
| Fume; Carbon monoxide; Carbon dioxide |
Chemical Identifiers
| 497-19-8 | |
| 105.99 | |
| CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
| 10340 | |
| disodium carbonate |
| CNa2O3 | |
| MFCD00003494 | |
| sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda | |
| CHEBI:29377 | |
| [Na+].[Na+].[O-]C([O-])=O |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature