missing translation for 'onlineSavingsMsg'
Learn More

Sodium Carbonate Anhydrous ACS, MilliporeSigma™

Catalog No. MSX03951
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MSX03951 500 g Poly Bottle
MSX03953 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MSX03951 Supplier Technidata Supplier No. SX03951
Only null left

Chemical Identifiers

CAS 497-19-8 , 7732-18-5
Molecular Formula CNa2O3
Molecular Weight (g/mol) 105.99
MDL Number MFCD00003494
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
Synonym sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
PubChem CID 10340
ChEBI CHEBI:29377
IUPAC Name disodium carbonate
SMILES [Na+].[Na+].[O-]C([O-])=O

Specifications

Physical Form Granules
Quantity 500 g
Linear Formula Na2CO3
Formula Weight 105.99
Grade ACS
Packaging Poly Bottle
Chemical Name or Material Sodium Carbonate, Anhydrous