missing translation for 'onlineSavingsMsg'
Learn More

Sodium Carbonate Andydrous Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014325
Change view
Click to view available options
Quantity:
2 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014325 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014325 Supplier Merck Emd Millipore Supplier No. PHR19482G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Specifications

CAS 497-19-8
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula Na2 CO3
MDL Number MFCD00003494
Melting Point 851°C
Synonym Sodium carbonate; Calcined soda; Carbonic acid disodium salt; Soda ash
Type Certified Reference Material
Quantity 2 g
Formula Weight 105.99
Grade Certified Reference Material