missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Carbonate, 1.00 N, Ricca Chemical
Supplier: Ricca Chemical Company 71901
Specifications
| Sodium Carbonate | |
| CNa2O3 | |
| sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda | |
| [Na+].[Na+].[O-]C([O-])=O | |
| 105.99 | |
| CHEBI:29377 | |
| 1.00N | |
| 11.5 to 12 |
| 497-19-8,7732-18-5 | |
| MFCD00003494 | |
| CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
| disodium carbonate | |
| 10340 | |
| Laboratory | |
| Natural Poly Bottle | |
| 4 L |
Chemical Identifiers
| 497-19-8 | |
| 105.99 | |
| CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
| 10340 | |
| disodium carbonate |
| CNa2O3 | |
| MFCD00003494 | |
| sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda | |
| CHEBI:29377 | |
| [Na+].[Na+].[O-]C([O-])=O |