missing translation for 'onlineSavingsMsg'
Learn More

Sodium Carbonate, 0.0454 N, 1 mL = 1 mg CO2, Ricca Chemical

Catalog No. 718016
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
718016 500 mL
718032 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 718016 Supplier Ricca Chemical Company Supplier No. 718016
Only null left

APHA for Carbon Dioxide Testing

Specifications

Chemical Name or Material Sodium Carbonate
Color Colorless
CAS 497-19-8, 7732-18-5
CAS Min % 98.0, 0.24
CAS Max % 100.0, 0.25
Molecular Formula CNa2O3
MDL Number MFCD00003494
Synonym sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
SMILES [Na+].[Na+].[O-]C([O-])=O
IUPAC Name disodium carbonate
Molecular Weight (g/mol) 105.99
PubChem CID 10340
Equivalence 1 mL = 1 mg CO
ChEBI CHEBI:29377
Grade Laboratory
Packaging Natural Poly Bottle
pH 11 to 11.5
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 0.0454Normal
Show More Show Less