missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPA for Alkalinity Testing
EPA for Alkalinity Testing
$81.59 - $190.72
Chemical Identifiers
| CAS | 497-19-8 |
|---|---|
| Molecular Formula | CNa2O3 |
| Molecular Weight (g/mol) | 105.99 |
| MDL Number | MFCD00003494 |
| InChI Key | CDBYLPFSWZWCQE-UHFFFAOYSA-L |
| Synonym | sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda |
| PubChem CID | 10340 |
| ChEBI | CHEBI:29377 |
| IUPAC Name | disodium carbonate |
| SMILES | [Na+].[Na+].[O-]C([O-])=O |
Chemical Identifiers
| 497-19-8 | |
| 105.99 | |
| CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
| 10340 | |
| disodium carbonate |
| CNa2O3 | |
| MFCD00003494 | |
| sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda | |
| CHEBI:29377 | |
| [Na+].[Na+].[O-]C([O-])=O |
Specifications
| Colorless | |
| 98.0,0.1 | |
| CNa2O3 | |
| sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda | |
| [Na+].[Na+].[O-]C([O-])=O | |
| 105.99 | |
| 10340 | |
| Laboratory | |
| 11 to 11.5 | |
| Liquid | |
| Sodium Carbonate |
| 497-19-8,7732-18-5 | |
| 100.0,0.11 | |
| MFCD00003494 | |
| CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
| disodium carbonate | |
| 1 mL = 1 mg CaCO | |
| CHEBI:29377 | |
| Natural Poly Bottle | |
| 120 mL | |
| 0.0200Normal |