missing translation for 'onlineSavingsMsg'
Learn More

Sodium Caprylate, 90%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor SO1451165LB

 View more versions of this product

Catalog No. 18613337


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1984-06-1
11.65 lb.
Reagent
1
90%
Poly Pail

Chemical Identifiers

1984-06-1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.