missing translation for 'onlineSavingsMsg'
Learn More

Sodium Caprylate, 90%, Spectrum™ Chemical

Catalog No. 18613335
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
1 kg
2.5 kg
11.65 lb.
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613335 10 g Poly Bottle
18613336 100 g Poly Bottle
18613341 500 g Poly Bottle
18613337 11.65 lb. Poly Pail
18613334 1 kg Poly Bottle
18613339 2.5 kg Poly Pail
18613338 12 kg Fiber Drum
18613340 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 18613335 Supplier Spectrum Chemical Mfg Cor Supplier No. SO14510GM

Chemical Identifiers

CAS 1984-06-1

Specifications

CAS Max % 1
Quantity 10 g
Percent Purity 90%
Grade Reagent
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.