missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Butyrate 98.0+%, TCI America™
Supplier: TCI America S0519100G
Specifications
| Sodium Butyrate | |
| 253°C | |
| C4H7NaO2 | |
| 100 g | |
| MFBOGIVSZKQAPD-UHFFFAOYSA-M | |
| sodium butanoate | |
| 5222465 | |
| 110.09 | |
| Crystalline Powder |
| 156-54-7 | |
| White | |
| MFCD00002816 | |
| sodium butyrate, sodium butanoate, butyric acid sodium salt, butanoic acid, sodium salt, butyrate sodium, butyric acid, sodium salt, sodium n-butyrate, sodium propanecarboxylate, butyric acid, na, butanoic acid, sodium salt 1:1 | |
| [Na+].CCCC([O-])=O | |
| 110.09 | |
| CHEBI:64103 | |
| ≥98.0% (T) |
Chemical Identifiers
| 156-54-7 | |
| 110.09 | |
| MFBOGIVSZKQAPD-UHFFFAOYSA-M | |
| 5222465 | |
| sodium butanoate |
| C4H7NaO2 | |
| MFCD00002816 | |
| sodium butyrate, sodium butanoate, butyric acid sodium salt, butanoic acid, sodium salt, butyrate sodium, butyric acid, sodium salt, sodium n-butyrate, sodium propanecarboxylate, butyric acid, na, butanoic acid, sodium salt 1:1 | |
| CHEBI:64103 | |
| [Na+].CCCC([O-])=O |
Safety and Handling
RTECSNumber : ET6400000
TSCA : Yes