missing translation for 'onlineSavingsMsg'
Learn More

Sodium Butyrate 98.0+%, TCI America™

Catalog No. p-7135917
Change view
Click to view available options
Quantity:
25 g
100 g
Catalog No. Quantity
S051925G 25 g
S0519100G 100 g
2 options

Catalog No. S051925G

Supplier: TCI America S051925G

Only null left

Chemical Identifiers

CAS 156-54-7
Molecular Formula C4H7NaO2
Molecular Weight (g/mol) 110.09
MDL Number MFCD00002816
InChI Key MFBOGIVSZKQAPD-UHFFFAOYSA-M
Synonym sodium butyrate, sodium butanoate, butyric acid sodium salt, butanoic acid, sodium salt, butyrate sodium, butyric acid, sodium salt, sodium n-butyrate, sodium propanecarboxylate, butyric acid, na, butanoic acid, sodium salt 1:1
PubChem CID 5222465
ChEBI CHEBI:64103
IUPAC Name sodium butanoate
SMILES [Na+].CCCC([O-])=O

Specifications

Melting Point 253°C
Color White
Quantity 25 g
Formula Weight 110.09
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Butyrate
RTECSNumber ET6400000
TSCA Yes