Learn More
Sodium butyrate, 98+%
CAS: 156-54-7 | C4H7NaO2 | 110.09 g/mol
Supplier: Thermo Scientific Chemicals A1107922
Description
Decreases calcium release from intracellular stores. Inhibits histone deacetylase.Sodium butyrate is used in the treatment of human spinal muscular atrophy patients. Further, it is used in the determination of the role of histone acetylation in chromatin structure and function.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDecreases calcium release from intracellular stores. Inhibits histone deacetylase.Sodium butyrate is used in the treatment of human spinal muscular atrophy patients. Further, it is used in the determination of the role of histone acetylation in chromatin structure and function.
Solubility
Soluble in water, dimethyl sulfoxide and ethanol. Insoluble in ether.
Notes
Incompatible with strong oxidizing agents.
Specifications
| Sodium butyrate | |
| >300°C | |
| C4H7NaO2 | |
| 100 g | |
| Hygroscopic | |
| Soluble in water,dimethyl sulfoxide and ethanol. Insoluble in ether. | |
| [Na+].CCCC([O-])=O | |
| 110.09 | |
| CHEBI:64103 |
| 156-54-7 | |
| >98% | |
| MFCD00002816 | |
| 3629439 | |
| sodium butyrate, sodium butanoate, butyric acid sodium salt, butanoic acid, sodium salt, butyrate sodium, butyric acid, sodium salt, sodium n-butyrate, sodium propanecarboxylate, butyric acid, na, butanoic acid, sodium salt 1:1 | |
| MFBOGIVSZKQAPD-UHFFFAOYSA-M | |
| sodium;butanoate | |
| 5222465 | |
| 110.09 |
Chemical Identifiers
| 156-54-7 | |
| 110.09 | |
| MFBOGIVSZKQAPD-UHFFFAOYSA-M | |
| 5222465 | |
| [Na+].CCCC([O-])=O |
| C4H7NaO2 | |
| MFCD00002816 | |
| sodium butyrate, sodium butanoate, butyric acid sodium salt, butanoic acid, sodium salt, butyrate sodium, butyric acid, sodium salt, sodium n-butyrate, sodium propanecarboxylate, butyric acid, na, butanoic acid, sodium salt 1:1 | |
| CHEBI:64103 |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 205-857-6
RTECSNumber : ET6400000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only