missing translation for 'onlineSavingsMsg'
Learn More

Sodium butyrate, 98+%

Catalog No. p-4407867
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1107906 5 g
AAA1107922 100 g
AAA1107936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1107906 Supplier Thermo Scientific Chemicals Supplier No. A1107906
Only null left

CAS: 156-54-7 | C4H7NaO2 | 110.09 g/mol

Decreases calcium release from intracellular stores. Inhibits histone deacetylase.Sodium butyrate is used in the treatment of human spinal muscular atrophy patients. Further, it is used in the determination of the role of histone acetylation in chromatin structure and function.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Decreases calcium release from intracellular stores. Inhibits histone deacetylase.Sodium butyrate is used in the treatment of human spinal muscular atrophy patients. Further, it is used in the determination of the role of histone ac

Chemical Identifiers

CAS 156-54-7
Molecular Formula C4H7NaO2
Molecular Weight (g/mol) 110.09
MDL Number MFCD00002816
InChI Key MFBOGIVSZKQAPD-UHFFFAOYSA-M
Synonym sodium butyrate, sodium butanoate, butyric acid sodium salt, butanoic acid, sodium salt, butyrate sodium, butyric acid, sodium salt, sodium n-butyrate, sodium propanecarboxylate, butyric acid, na, butanoic acid, sodium salt 1:1
PubChem CID 5222465
ChEBI CHEBI:64103
SMILES [Na+].CCCC([O-])=O

Specifications

Melting Point >300°C
Linear Formula CH3(CH2)2CO2Na
Quantity 5 g
Beilstein 3629439
Sensitivity Hygroscopic
Solubility Information Soluble in water,dimethyl sulfoxide and ethanol. Insoluble in ether.
IUPAC Name sodium butanoate
Formula Weight 110.09
Percent Purity ≥98%
Chemical Name or Material Sodium butyrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335;
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 205-857-6
RTECSNumber ET6400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only