missing translation for 'onlineSavingsMsg'
Learn More

Sodium bromate, 99+%, extra pure

Catalog No. p-221936
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC208871000 100 g Glass bottle
AC208875000 500 g Glass bottle
AC208870025 2.5 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC208871000 Supplier Thermo Scientific Chemicals Supplier No. 208871000

CAS: 7789-38-0 | BrNaO3 | 150.9 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent

Chemical Identifiers

CAS 7789-38-0
Molecular Formula BrNaO3
Molecular Weight (g/mol) 150.9
MDL Number MFCD00003476
InChI Key XUXNAKZDHHEHPC-UHFFFAOYSA-M
Synonym sodium bromate, bromic acid, sodium salt, dyetone, bromate de sodium, sodium bromate dot, neutralizer k-126, neutralizer k-140, neutralizer k-938, sodium bromate nabro3, bromate de sodium french
PubChem CID 23668195
ChEBI CHEBI:75229
IUPAC Name sodium;bromate
SMILES [O-]Br(=O)=O.[Na+]

Specifications

Melting Point 381.0°C
Color White
Physical Form Fine Crystalline Powder
Quantity 100 g
Assay Percent Range 99+%
Linear Formula NaBrO3
Fieser 01,1055
Merck Index 15,8728
Solubility Information Solubility in water: 364g/L (20°C). Other solubilities: insoluble in alcohol , ether and aceton
Formula Weight 150.9
Percent Purity ≥99%
Grade Extra Pure
Packaging Glass bottle
Specific Gravity 3.33
Density 3.3300g/mL
Chemical Name or Material Sodium bromate, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May intensify fire; oxidizer.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse caut
EINECSNumber 232-160-4
RTECSNumber EF8750000
TSCA TSCA

RUO – Research Use Only