missing translation for 'onlineSavingsMsg'
Learn More

Sodium bromate, 99.5% (metals basis)

CAS: 7789-38-0 | BrNaO3 | 150.891 g/mol

$65.15 - $407.78

Chemical Identifiers

CAS 7789-38-0
Molecular Formula BrNaO3
Molecular Weight (g/mol) 150.891
MDL Number MFCD00003476
InChI Key XUXNAKZDHHEHPC-UHFFFAOYSA-M
Synonym sodium bromate, bromic acid, sodium salt, dyetone, bromate de sodium, sodium bromate dot, neutralizer k-126, neutralizer k-140, neutralizer k-938, sodium bromate nabro3, bromate de sodium french
PubChem CID 23668195
ChEBI CHEBI:75229
IUPAC Name sodium;bromate
SMILES [O-]Br(=O)=O.[Na+]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1410122
View Documents
Thermo Scientific Chemicals
01410122
100 g
Each for $65.15
Only null left
 
AA1410136
View Documents
Thermo Scientific Chemicals
01410136
500 g
Each for $167.70
Only null left
 
AA14101A3
View Documents
Thermo Scientific Chemicals
014101A3
2 kg
Each for $407.78
Only null left
 
Description

Description

Sodium bromate is used as an oxidant to convert tetrahydropyranyl ethers to carbonyl compounds. It acts as an analytical reagent. It is also used for dissolving gold from its ores, boiler cleaning, oxidation of sulfur and vat dyes. Further, it is used in hair treatment formulations as a hair-permanent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7789-38-0
150.891
XUXNAKZDHHEHPC-UHFFFAOYSA-M
23668195
sodium;bromate
BrNaO3
MFCD00003476
sodium bromate, bromic acid, sodium salt, dyetone, bromate de sodium, sodium bromate dot, neutralizer k-126, neutralizer k-140, neutralizer k-938, sodium bromate nabro3, bromate de sodium french
CHEBI:75229
[O-]Br(=O)=O.[Na+]

Specifications

381°C (decomposition)
White
100 g
BrNaO3
sodium bromate, bromic acid, sodium salt, dyetone, bromate de sodium, sodium bromate dot, neutralizer k-126, neutralizer k-140, neutralizer k-938, sodium bromate nabro3, bromate de sodium french
XUXNAKZDHHEHPC-UHFFFAOYSA-M
sodium;bromate
23668195
150.9
1.594
3.34 g/cm3
7789-38-0
Crystals
99.5% (metals basis)
MFCD00003476
Soluble in water (2.5 parts in cold water,1.1 part in boiling water)Soluble in water and ammonia. Insoluble in ethanol,acetone and ether.
[O-]Br(=O)=O.[Na+]
150.891
CHEBI:75229
Odorless
White crystalline
Sodium bromate
Videos
Safety and Handling

Safety and Handling

P210-P220-P221-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c

H272-H302-H315-H319-H335

DOTInformation : Hazard Class: 5.1, Packing Group: II

EINECSNumber : 232-160-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only