missing translation for 'onlineSavingsMsg'
Learn More

Sodium Borohydride 95.0+%, TCI America™

Catalog No. S048025G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S048025G 25 g
S0480100G 100 g
S0480500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S048025G Supplier TCI America Supplier No. S048025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16940-66-2
Molecular Formula BH4Na
Molecular Weight (g/mol) 37.83
MDL Number MFCD00003518
InChI Key YOQDYZUWIQVZSF-UHFFFAOYSA-N
Synonym Sodium Tetrahydroborate, Sodium Tetrahydridoborate
IUPAC Name sodium boranuide
SMILES [BH4-].[Na+]

Specifications

Color White
Physical Form Crystalline Powder
Quantity 25 g
UN Number 1426
Formula Weight 37.83
Percent Purity ≥95.0% (T)
Chemical Name or Material Sodium Borohydride
EINECSNumber (1)-0061
RTECSNumber ED3325000
TSCA Yes