missing translation for 'onlineSavingsMsg'
Learn More

Sodium Borohydride (Powder), Fisher Chemical™

Catalog No. S67825
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S67825 25 g Poly Bottle
S67810 10 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S67825 Supplier Thermo Fisher Scientific Supplier No. S678 25
Only null left

Chemical Identifiers

CAS 16940-66-2
Molecular Formula BH4Na
Molecular Weight (g/mol) 37.83
MDL Number MFCD00003518
InChI Key YOQDYZUWIQVZSF-UHFFFAOYSA-N
IUPAC Name sodium boranuide
SMILES [BH4-].[Na+]

Specifications

Melting Point 360°C
Color White
pH 11
Physical Form Solid
Quantity 25 g
Formula Weight 37.83
Identification Pass Test
Packaging Poly Bottle
Vapor Pressure Negligible
Decomposition Information >220°C
Chemical Name or Material Sodium Borohydride
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Water reactive. Toxic by inhalation, in contact with skin and if swallowed. Causes burns by all exposure routes. Hygroscopic. Use personal protective equipment. Do not allow contact with water. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:3
Flammability:1
Instability:2
DOTInformation DOT Class 4.3, : Dangerous When Wet