missing translation for 'onlineSavingsMsg'
Learn More

Sodium Borohydride, Crystalline Powder, Reagent, ACS, 98%, Spectrum™ Chemical

Catalog No. 18606368
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
40 kg
50 kg
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606368 2.5 kg Amber Glass Bottle
18606365 25 g Amber Glass Bottle
18606366 100 g Amber Glass Bottle
18606367 500 g Amber Glass Bottle
18606370 40 kg Amber Glass Bottle
18606369 50 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18606368 Supplier Spectrum Chemical Mfg Cor Supplier No. S11872.5KG
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

S1187, 16940-66-2, NaBH4

Spectrum™ Chemical Sodium Borohydride, Crystalline Powder, Reagent, ACS is an inorganic compound that is white in color and usually found as a powder. It is a reducing agent with versatility and is used as a synthetic reagent in the laboratory. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 16940-66-2
Molecular Formula BH4Na
Molecular Weight (g/mol) 37.83
MDL Number MFCD00003518
InChI Key YOQDYZUWIQVZSF-UHFFFAOYSA-N
IUPAC Name sodium boranuide
SMILES [BH4-].[Na+]

Specifications

CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 2.5 kg
Percent Purity 98%
Grade Reagent
Packaging Amber Glass Bottle
Chemical Name or Material Sodium Borohydride, Crystalline Powder