missing translation for 'onlineSavingsMsg'
Learn More

Sodium borohydride, 99%, (VenPureTM SF granules), granules, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 448500100

Catalog No. AC448500100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Sodium borohydride
10kg
H4BNa
15,8727
Solubility in water: reacts. Other solubilities: soluble in ammonia- reacts with acids,soluble in amines,pyridine and dimethylether of,diethylene glycol,insol.in other ethers,hydrocarb.,alkyl chlorides,reacts with oxidizing agents
37.83
Plastic drum
16940-66-2
98.5% min. (Hydrogen evolution)
NaBH4
Sodium tetrahydroborate, SBH
37.83
99%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Toxic if swallowed.
May damage fertility.
May damage the unborn child.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts viol

GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Keep away from any possible contact with water,because of violent reaction and possible flash fire.
IF INHALED: Remove to fresh air and keep at rest in a position comf

GHS Signal Word: Danger

EINECSNumber : 241-004-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only