Learn More
Sodium borohydride, 99%, (VenPureTM SF powder), powder
CAS: 16940-66-2 | BH4Na | 37.83 g/mol
$186.85 - $186.85
Chemical Identifiers
| CAS | 16940-66-2 |
|---|---|
| Molecular Formula | BH4Na |
| Molecular Weight (g/mol) | 37.83 |
| MDL Number | MFCD00003518 |
| InChI Key | YOQDYZUWIQVZSF-UHFFFAOYSA-N |
| Synonym | Sodium tetrahydroborate, SBH |
| IUPAC Name | sodium boranuide |
| SMILES | [BH4-].[Na+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC448481000
|
Thermo Scientific Chemicals
448481000 |
100 g |
Each for $186.85
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 16940-66-2 | |
| 37.83 | |
| YOQDYZUWIQVZSF-UHFFFAOYSA-N | |
| sodium boranuide |
| BH4Na | |
| MFCD00003518 | |
| Sodium tetrahydroborate, SBH | |
| [BH4-].[Na+] |
Specifications
| 360.0°C | |
| 100 g | |
| BH4Na | |
| MFCD00003518 | |
| Sodium tetrahydroborate, SBH | |
| YOQDYZUWIQVZSF-UHFFFAOYSA-N | |
| sodium boranuide | |
| 37.83 | |
| Plastic Bottle |
| 16940-66-2 | |
| 99% | |
| NaBH4 | |
| 15,8727 | |
| Solubility in water: reacts. Other solubilities: soluble in ammonia - reacts with acids, soluble in amines, pyridine and dimethylether of, diethylene glycol, insol.in other ethers, hydrocarb., alkyl chlorides, reacts with oxidizing agents | |
| [BH4-].[Na+] | |
| 37.83 | |
| 99% | |
| Sodium borohydride |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Toxic if swallowed.
May damage fertility.
May damage the unborn child.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts viol
GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Keep away from any possible contact with water,because of violent reaction and possible flash fire.
IF INHALED: Remove to fresh air and keep at rest in a position comf
GHS Signal Word: Danger
EINECSNumber : 241-004-4
RUO – Research Use Only