missing translation for 'onlineSavingsMsg'
Learn More

Sodium borohydride, 99%, (VenPureTM AF caplets), caplets

CAS: 16940-66-2 | BH4Na | 37.83 g/mol

$570.69 - $570.69

Chemical Identifiers

CAS 16940-66-2
Molecular Formula BH4Na
Molecular Weight (g/mol) 37.83
MDL Number MFCD00003518
InChI Key YOQDYZUWIQVZSF-UHFFFAOYSA-N
Synonym Sodium tetrahydroborate, SBH
IUPAC Name sodium boranuide
SMILES [BH4-].[Na+]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC448495000
SDS View Documents
Thermo Scientific Chemicals
448495000
500 g
Each for $570.69
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

16940-66-2
37.83
YOQDYZUWIQVZSF-UHFFFAOYSA-N
sodium boranuide
BH4Na
MFCD00003518
Sodium tetrahydroborate, SBH
[BH4-].[Na+]

Specifications

360.0°C
500 g
BH4Na
MFCD00003518
Sodium tetrahydroborate, SBH
YOQDYZUWIQVZSF-UHFFFAOYSA-N
sodium boranuide
37.83
Plastic Bottle
16940-66-2
99%
NaBH4
15,8727
Solubility in water: reacts. Other solubilities: soluble in ammonia - reacts with acids, soluble in amines, pyridine and dimethylether of, diethylene glycol, insol.in other ethers, hydrocarb., alkyl chlorides, reacts with oxidizing agents
[BH4-].[Na+]
37.83
99%
Sodium borohydride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Toxic if swallowed.
May damage fertility.
May damage the unborn child.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts viol

GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Keep away from any possible contact with water,because of violent reaction and possible flash fire.
IF INHALED: Remove to fresh air and keep at rest in a position comf

GHS Signal Word: Danger

EINECSNumber : 241-004-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only