missing translation for 'onlineSavingsMsg'
Learn More

Sodium Borohydride, ≥99%, Honeywell Fluka™

Catalog No. p-7120770
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60047350 25 g Glass bottle
60047349 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60047350 Supplier Honeywell Chemicals Supplier No. 7132125G
Only null left

TraceSELECT™, for the determination of hydride formers by AAS,≥99%

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions

 


 

Chemical Identifiers

CAS 16940-66-2
Molecular Formula BH4Na
Molecular Weight (g/mol) 37.83
MDL Number MFCD00003518
InChI Key YOQDYZUWIQVZSF-UHFFFAOYSA-N
IUPAC Name sodium boranuide
SMILES [BH4-].[Na+]

Specifications

Boiling Point >400°C
Melting Point >360°C
Color White
Physical Form Prill or Powder
Quantity 25 g
Linear Formula NaBH4
UN Number UN1426
Formula Weight 37.83g/mol
Percent Purity ≥99%
Odor Odorless
Packaging Glass bottle
Vapor Pressure >0.000054 Pa
Density 1.074g/cm3
Chemical Name or Material Sodium borohydride
Show More Show Less
Hazard Statement P201-P231 + P232-P280-P308 + P313-P370 + P378-P402 + P404
DOTInformation 4.3 / PGI
RTECSNumber 241-004-4