missing translation for 'onlineSavingsMsg'
Learn More

Sodium borohydride, 98+%, powder

Catalog No. p-41817
Change view
Click to view available options
Quantity:
5 g
10 g
Packaging:
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC189300050 5 g Plastic Bottle
AC189300100 10 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC189300050 Supplier Thermo Scientific Chemicals Supplier No. 189300050
Only null left

CAS: 16940-66-2 | BH4Na | 37.83 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent

Chemical Identifiers

CAS 16940-66-2
Molecular Formula BH4Na
Molecular Weight (g/mol) 37.83
MDL Number MFCD00003518
InChI Key YOQDYZUWIQVZSF-UHFFFAOYSA-N
Synonym Sodium tetrahydroborate, SBH
IUPAC Name sodium boranuide
SMILES [BH4-].[Na+]

Specifications

Melting Point 360.0°C
Color White
Physical Form Powder
Quantity 5 g
Assay Percent Range 98% min. (Iodometry)
Linear Formula NaBH4
Fieser 01,1049; 02,377; 03,262; 04,443; 05,597; 06,530; 07,329; 08,449; 09,420; 10,357; 11,477; 12,441; 13,21; 14,99; 15,290; 16,304; 17,189
Merck Index 15, 8727
Solubility Information Solubility in water: reacts. Other solubilities: soluble in ammonia - reacts with acids, soluble in amines, pyridine and dimethylether of, diethylene glycol, insol.in other ethers, hydrocarb., alkyl chlorides, reacts with oxidizing agents
Formula Weight 37.83
Percent Purity ≥98%
Packaging Plastic Bottle
Vapor Pressure Negligible
Decomposition Information >220°C
Density 1.3
Chemical Name or Material Sodium borohydride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Toxic if swallowed.
May damage fertility.
May damage the unborn child.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts viol
Precautionary Statement GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Keep away from any possible contact with water,because of violent reaction and possible flash fire.
IF INHALED: Remove to fresh air and keep at rest in a position comf
EINECSNumber 241-004-4
RTECSNumber ED3325000
TSCA TSCA

RUO – Research Use Only