missing translation for 'onlineSavingsMsg'
Learn More

Sodium borohydride, 0.5M solution in diglyme, AcroSeal™

Catalog No. AC191131000
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC191131000 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC191131000 Supplier Thermo Scientific Chemicals Supplier No. 191131000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16940-66-2 | BH4Na | 37.83 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent

Specifications

Chemical Name or Material Sodium borohydride
Color Colorless
Name Note 0.5M Solution in Diglyme
CAS 16940-66-2, 111-96-6
Concentration 0.45 to 0.55M
Density 0.9450g/mL
Packaging AcroSeal™ Glass bottle
Specific Gravity 0.945
Molecular Formula BH4Na
Linear Formula NaBH4
Quantity 100 mL
MDL Number MFCD00003518
Flash Point 69°C
Fieser 01,1049; 02,377; 03,262; 04,443; 05,597; 06,530; 07,329; 08,449; 09,420; 10,357; 11,477; 12,441; 13,21; 14,99; 15,290; 16,304; 17,189
Physical Form Liquid
Merck Index 15, 8727
Synonym Sodium tetrahydroborate, SBH
Solubility Information Solubility in water: soluble in cold water, reacts with hot water.
InChI Key YOQDYZUWIQVZSF-UHFFFAOYSA-N
SMILES [BH4-].[Na+]
IUPAC Name sodium boranuide
Molecular Weight (g/mol) 37.83
Formula Weight 37.82
Show More Show Less

RUO – Research Use Only