missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Borate, Decahydrate, NF, 99-105%, Spectrum™ Chemical
Description
Spectrum™ Chemical Sodium Borate, Decahydrate, NF is an anti-fungal agent in pharmaceutical formulations. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 1 |
| Quantity | 500 g |
| Percent Purity | 99 to 105% |
| Grade | NF |
| Packaging | Poly Bottle |