missing translation for 'onlineSavingsMsg'
Learn More

Sodium Borate, Decahydrate, NF, 99-105%, Spectrum™ Chemical

Catalog No. 18606362
Change view
Click to view available options
Quantity:
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606362 2.5 kg Poly Bottle
18606363 12 kg Poly Pail
18606364 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18606362 Supplier Spectrum Chemical Mfg Cor Supplier No. S11832.5KG
Only null left

S1183, 1303-96-4, Na2B4O7·10H2O

Spectrum™ Chemical Sodium Borate, Decahydrate, NF is an anti-fungal agent in pharmaceutical formulations. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1303-96-4
Molecular Formula B4H20Na2O17
Molecular Weight (g/mol) 381.36
InChI Key OJPFLODMGUZMKD-UHFFFAOYSA-N
IUPAC Name disodium decahydrate oxoboranyl ({oxido[(oxoboranyl)oxy]boranyl}oxy)boronate
SMILES O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]B(OB=O)OB([O-])OB=O

Specifications

CAS Max % 1
Quantity 2.5 kg
Percent Purity 99 to 105%
Grade NF
Packaging Poly Bottle