missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Borate, Decahydrate, ACS Grade, 99.5 to 105.0%, LabChem™
Supplier: LabChem LC229551
Specifications
| 320°C | |
| 75°C | |
| 1303-96-4 | |
| 100 | |
| 500 g | |
| Borax; Sodium Tetraborate | |
| 381.37 | |
| ACS | |
| Poly Bottle | |
| Fume; Sodium oxide; Boron |
| Sodium Borate | |
| Decahydrate | |
| White | |
| Powder | |
| Na2B4O7·10H2O | |
| Soluble in water | |
| 99.5 to 105.0% | |
| Passes Test | |
| 1.73g/cm3 | |
| 1.73g/cm3 |
Chemical Identifiers
| 1303-96-4 | |
| Borax; Sodium Tetraborate |
Safety and Handling
GHS H Statement
May damage fertility or the unborn child.
GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear eye protection, protective gloves.
If exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
EINECSNumber : 215-540-4
Recommended Storage : Room Temperature