missing translation for 'onlineSavingsMsg'
Learn More

Sodium Borate Decahydrate ACS, MilliporeSigma™

Catalog No. MSX03551
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MSX03551 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MSX03551 Supplier Technidata Supplier No. SX03551

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ACS reagent grade sodium borate, decahydrate for laboratory applications.

Chemical Identifiers

CAS 1303-96-4

Specifications

Physical Form Crystals
Quantity 500 g
Linear Formula Na2B4O7·10H2O
Formula Weight 381.37
Percent Purity 99.5 to 105.0%
Grade ACS
Packaging Poly Bottle
Chemical Name or Material Sodium Borate, Decahydrate