missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sodium borate, 0.5M buffer soln., pH 9.0

Catalog No. p-7042751
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63637AK 250 mL
AAJ63637AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63637AK Supplier Thermo Scientific Chemicals Supplier No. J63637AK
Only null left

CAS: 1330-43-4 | B4Na2O7 | 201.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Sodium borate is used as anhydrous borax for proteomics research, used in the manufacturing of glazes and enamels, a component in tanning agents; curing and preserving skins, a agent used in cleaning compounds, used to artificially age wood, as a preservative, either alone or with other antiseptics against wood fungus, used in fireproofing fabrics and wood, used as antiseptic, used in detergents, an astringent for mucous membranes.

Solubility
Fully miscible.

Notes
Protect from heat. Store away from oxidizing agents.

Chemical Identifiers

CAS 1330-43-4
Molecular Formula B4Na2O7
Molecular Weight (g/mol) 201.21
MDL Number MFCD00081185
InChI Key NXSVMNYRVBQCMO-UHFFFAOYSA-N
IUPAC Name disodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate)
SMILES [Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2

Specifications

Color Colorless
Concentration 0.5 M
pH 9.0
Physical Form Liquid
Quantity 250 mL
Solubility Information Fully miscible.
Chemical Name or Material Sodium borate buffer soln.
Hazard Category H319-H360FD
Hazard Statement GHS H Statement
H360
May damage fertility or the unborn child.
Precautionary Statement P201-P202-P264b-P280i-P281-P305+P351+P338-P308+P313-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only