missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sodium borate, 0.5M buffer soln., pH 8.0

Catalog No. p-7041738
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60068AK 250 mL
AAJ60068AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60068AK Supplier Thermo Scientific Chemicals Supplier No. J60068AK
Only null left

CAS: 1330-43-4 | B4Na2O7 | 201.21 g/mol

It is a component of many detergents, cosmetics, and enamel glazes. It is also used to make buffer solutions in biochemistry, as a fire retardant, as an anti-fungal compound for fiberglass, as a flux in metallurgy, neutron-capture shields for radioactive sources, a texturing agent in cooking, and as a precursor for other boron compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a component of many detergents, cosmetics, and enamel glazes. It is also used to make buffer solutions in biochemistry, as a fire retardant, as an anti-fungal compound for fiberglass, as a flux in metallurgy, neutron-capture shields for radioactive sources, a texturing agent in cooking, and as a precursor for other boron compounds.

Solubility
Fully miscible with water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 1330-43-4
Molecular Formula B4Na2O7
Molecular Weight (g/mol) 201.21
MDL Number MFCD00081185
InChI Key NXSVMNYRVBQCMO-UHFFFAOYSA-N
IUPAC Name disodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate)
SMILES [Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2

Specifications

Concentration 0.5 M
pH 8.0
Physical Form Liquid
Quantity 250 mL
Solubility Information Fully miscible with water.
Chemical Name or Material Sodium borate buffer soln.
Hazard Category H319-H360FD
Hazard Statement GHS H Statement
H360
May damage fertility or the unborn child.
Precautionary Statement P201-P202-P264b-P280i-P281-P305+P351+P338-P308+P313-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only