missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bitartrate, Monohydrate, Powder, Reagent, 99%, Spectrum™ Chemical

Catalog No. p-7158188
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606347 125 g Poly Bottle
18606348 500 g Poly Bottle
18606349 2.5 kg Poly Bottle
18606350 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18606347 Supplier Spectrum Chemical Mfg Cor Supplier No. S1175125GM
Only null left

S1175, 6131-98-2, NaHC4H4O6·H2O

Spectrum™ Chemical Sodium Bitartrate, Monohydrate, Powder, Reagent The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 6131-98-2
Molecular Formula C4H7NaO7
Molecular Weight (g/mol) 190.08
InChI Key LLVQEXSQFBTIRD-UHFFFAOYNA-M
IUPAC Name sodium 3-carboxy-2,3-dihydroxypropanoate hydrate
SMILES O.[Na+].OC(C(O)C([O-])=O)C(O)=O

Specifications

CAS Max % 1
Quantity 125 g
Percent Purity 99%
Grade Reagent
Packaging Poly Bottle